missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-3,5-DHPG
GREENER_CHOICE

Product Code. 18727536
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18727536 10 mg 10mg
18745936 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18727536 Supplier Tocris Bioscience™ Supplier No. 0805/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective group I mGlu agonist; active enantiomer of 3,5-DHPG (Cat. No. 0342)

(S)-3,5-DHPG is a selective group I mGlu receptor agonist. (S)-3,5-DHPG regulates the expression of microRNAs in mouse cerebral cortex in vivo. (S)-3,5-DHPG induces a profound depression of synaptic dopamine release in mouse cortico-striatal cells in vitro, and anxiety-like behavior in vivo in mice.

Racemate also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162870-29-3
Molecular Formula C8H9NO4

Specifications

Quantity 10 mg
Formula Weight 183.16
Purity >98%
Molecular Weight (g/mol) 183.16
Chemical Name or Material (S)-3, 5-DHPG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.