missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-5-methylhexanoic acid, 95%

Product Code. 15478717
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15478717

Brand: Thermo Scientific Alfa Aesar H52173.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132549-43-0 | C12H23NO4 | 245.32 g/mol

(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of carba peptides and building block for β-peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of ″carba peptides″ and building block for β-peptides.

Solubility
Very slightly soluble (1.1 g/L at 25°C).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132549-43-0
Molecular Formula C12H23NO4
Molecular Weight (g/mol) 245.32
MDL Number MFCD02101665
InChI Key XRVAMBSTOWHUMM-VIFPVBQESA-N
Synonym boc-l-beta-homoleucine, s-3-tert-butoxycarbonyl amino-5-methylhexanoic acid, boc-beta-homoleu-oh, boc-l-beta-hleu-oh, hexanoic acid, 3-1,1-dimethylethoxy carbonyl amino-5-methyl-, 3s, 3s-3-tert-butoxycarbonyl amino-5-methylhexanoic acid, ambotzbaa6190, boc-leu-c*ch2 oh, pubchem12105, rarechem em wb 0131
PubChem CID 2761526
IUPAC Name (3S)-5-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)C[C@@H](CC(O)=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 53°C to 55°C
Quantity 1 g
Beilstein 4251252
Solubility Information Very slightly soluble (1.1g/L at 25°C).
Formula Weight 245.32
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-5-methylhexanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.