missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S-arrestin Monoclonal Antibody (S128)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547477
This item is not returnable.
View return policy
Description
S-arrestin Monoclonal Antibody for Western Blot, ICC/IF, IHC
S-arrestin, also known as retinal arrestin or visual arrestin, is part of a small gene family that includes four members. It primarily functions in the retina, specifically within rod and cone photoreceptors. S-arrestin plays a crucial role in phototransduction by participating in the inactivation of opsin, which is a key component in the visual signaling pathway. Upon light stimulation, activated opsin interacts with a G-protein-coupled receptor kinase (GRK), leading to its phosphorylation. S-arrestin then binds to phosphorylated opsin to terminate the G-protein signaling, hence contributing to the process of vision adaptation and regulation of neurotransmitter release at the photoreceptor-bipolar cell synapse. This interaction is vital for the proper functioning of the visual cycle, ensuring that 11-cis-retinal is regenerated through the retinoid cycle. Mutations in the S-arrestin gene, like other rhodopsin alleles, are linked to various retinal disorders. The gene's expression pattern is specific to the retina, underscoring its role in vision, and it is distinct from other members of the arrestin family like beta-arrestins which regulate G-protein-coupled receptors (GPCRs) outside of the retina.
Specifications
| S-arrestin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P08168, P10523, P15887, P20443, P79260 | |
| SAG | |
| Recombinant bovine arrestin 1 with the first 20 amino acids of the C-terminus truncated. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig, Horse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| S128 | |
| Unconjugated | |
| SAG | |
| 48 kDa protein; A930001K18Rik; Arr1; arrestin; arrestin 1; arrestin1; arrestin-1; DKFZp686D1084; DKFZp686I1383; Irbp; photoreceptor protein/alleged cGMP phosphodiesterase activation modulator; put. S-antigen C-terminus; retinal S antigen; retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; rod photoreceptor arrestin; RP47; Sag; S-AG; SAGMR; SANTI; S-antigen; S-antigen visual arrestin; S-antigen, retina and pineal gland (arrestin); S-antigen; retina and pineal gland (arrestin); S-arrestin; S-arrestin short form; visual arrestin 1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100064951, 20215, 25539, 280922, 397151, 6295 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction