missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8/S100A9 Heterodimer Antibody (5.5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 372.00
Specifications
| Antigen | S100A8/S100A9 Heterodimer |
|---|---|
| Clone | 5.5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607906
|
Novus Biologicals
NBP2-50283-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676136
|
Novus Biologicals
NBP2-50283 |
0.1 mL |
€ 372.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A8/S100A9 Heterodimer Monoclonal antibody specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOFSpecifications
| S100A8/S100A9 Heterodimer | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS | |
| IgG1 | |
| Protein A purified |
| 5.5 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Acute monocytic leukaemia cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title