missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100B Mouse, Clone: SH-B1, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6285-MSM6-P0
This item is not returnable.
View return policy
Specifications
| S100B | |
| Cytoplasmic and Nuclear | |
| SH-B1 | |
| Unconjugated | |
| XP_016883913 | |
| Mouse | |
| 10-12kDa | |
| Protein A/G Purified | |
| RUO | |
| 6285 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| S100B | |
| Purified bovine brain S100 protein | |
| Brain, Melanoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction