Learn More
S100B Mouse, Clone: SH-B1, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | S100B |
|---|---|
| Clone | SH-B1 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15960848
|
EnquireBio
6285-MSM6-P0 |
20 μg | |||||||
|
15970848
|
EnquireBio
6285-MSM6-P1 |
100 μg |
€ 554.40 € 478.00 / 100µg Save € 76.40 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| S100B | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 6285 | |
| Purified bovine brain S100 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 10-12kDa |
| SH-B1 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| XP_016883913 | |
| S100B | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic and Nuclear | |
| Brain, Melanoma |