Learn More
Hirschmann™ Safe-Cone Filters for Labopette™
For the prevention of cross contamination of liquid samples or vapours when pipetting radioactive, biohazardous or corrosive samples.
Brand: Hirschmann™ 9470011
Additional Details : Weight : 0.25000kg
Description
- Provides the decisive advantage when protecting pipettes and samples against contamination
- Extends the service life of pipettes
- Provides excellent protection for the internal components if replaced regularly
- Can serve as additional protection against penetration of liquids into the interior of the pipette
- For use with a range of Labopette™ models
Specifications
Safety filter | |
Protection against cross-contamination | |
Labopette models 9480104, 9480105, 9480106, 9475409, 9475508, 9475509, 9475510, 9480800 |