Learn More
Hirschmann™ Safe-Cone Filters for Labopette™
For the prevention of cross contamination of liquid samples or vapours when pipetting radioactive, biohazardous or corrosive samples.
Brand: Hirschmann™ 9470010
Additional Details : Weight : 0.25000kg
Description
- Provides the decisive advantage when protecting pipettes and samples against contamination
- Extends the service life of pipettes
- Provides excellent protection for the internal components if replaced regularly
- Can serve as additional protection against penetration of liquids into the interior of the pipette
- For use with a range of Labopette™ models
Specifications
Safety filter | |
Protection against cross-contamination | |
Labopette models 9475404, 9475405, 9475511, 9475512, 9475513 |