missing translation for 'onlineSavingsMsg'
Learn More

Saikogenin A, MedChemExpress

Product Code. 16441758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441758 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441758 Supplier Medchem Express Supplier No. HYN6584

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Saikogenin A, extracted from a Chinese herbal plant called Tsai-Fu, is a dipeptidyl peptidase-IV (DPP-IV) inhibitor.

Saikogenin A, extracted from a Chinese herbal plant called Tsai-Fu, is a dipeptidyl peptidase-IV (DPP-IV) inhibitor.

Specifications

Chemical Name or Material Saikogenin A
CAS 5092-09-1
Color White
Purity Grade Notes Research
Molecular Formula C30H48O4
Quantity 5 mg
SMILES C[C@]12[C@]3(C(C=C[C@]1([H])[C@@]4([C@@]([C@@](C)([C@@H](O)CC4)CO)([H])CC2)C)=C5[C@](CO)(CCC(C)(C)C5)[C@@H](O)C3)C
Molecular Weight (g/mol) 472.7
Formula Weight 472.7
Percent Purity 98.31%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.