missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12586-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.Specifications
| SAMHD1 | |
| Polyclonal | |
| Unconjugated | |
| SAMHD1 | |
| DCIP, HDDC1, Monocyte protein 5, MOP 5, MOP5, SAM domain and HD domain 1, SAMHD1, SBBI88 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25939, 311580, 56045 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q60710, Q9Y3Z3 | |
| Samhd1 | |
| SAMHD1 Fusion Protein Ag3287 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction