missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | SAMHD1 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16810374
|
Proteintech
12586-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16800374
|
Proteintech
12586-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.Specifications
| SAMHD1 | |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q60710, Q9Y3Z3 | |
| 25939, 311580, 56045 | |
| SAMHD1 Fusion Protein Ag3287 | |
| Primary | |
| -20°C | |
| SAMHD1 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DCIP, HDDC1, Monocyte protein 5, MOP 5, MOP5, SAM domain and HD domain 1, SAMHD1, SBBI88 | |
| Samhd1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title