Learn More
Skytec™ Sapphire Aero PU Gloves
Product Code. p-200110068
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274586
Brand: Skytec™ SKG00044FD
Only null left
Add to basket
Add to basket
A cut resistant glove giving a 'second skin' feel, thin, soft and comfortable allowing high dexterity tasks. PU palm coating to help handling small parts.
  • Ultra Lightweight Glass Fiber Free Glove Offering Medium Level Cut Resistance with a PU Coated Grip Palm
  • Medium cut resistance: Level C / ANSI A3
  • 18 gauge seamless liner offers flexibilty
  • Touchscreen capabilities to allow usage with screen devices
  • Designed for easy movement and continuous wear
  • Breathable back of hand to reduce perspiration
  • Seamless knit prevents irritation
  • Elasticated wrist provides a secure fit
Specifications
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A3, EN 388:2016 4X42C, EN 388:2016 + A1:2018 3X42C, EN 21420:2020
Coating Material Polyurethane
Color Gray, Black
Cut Resistance Rating Level C
Hand Ambidextrous
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size Small
EN388 Standard 3X42C
Length (Metric) 240 mm