Learn More
SARS-CoV-2 Spike Protein Monoclonal Antibody (OTI4C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814463
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 Spike Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein in Virus samples. It is validated for ELISA, Western BlotSpecifications
| SARS-CoV-2 Spike Protein | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| OTI4C6 | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli. | |
| 100 μL | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.