Learn More
SARS-CoV-2 Spike Protein Monoclonal Antibody (OTI4C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | SARS-CoV-2 Spike Protein |
|---|---|
| Clone | OTI4C6 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17569214
|
Origene Technologies
TA814463S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17549214
|
Origene Technologies
TA814463 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 Spike Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein in Virus samples. It is validated for ELISA, Western BlotSpecifications
| SARS-CoV-2 Spike Protein | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles |
| OTI4C6 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| IgG2a | |
| Protein A/G | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.