Learn More
Sarsasapogenin, MedChemExpress
Description
Sarsasapogenin is a sapogenin from the Chinese medical herb Anemarrhena asphodeloides Bunge, with antidiabetic, anti-oxidative, anticancer and anti-inflamatory activities.
Specifications
Specifications
| Chemical Name or Material | Sarsasapogenin |
| CAS | 126-19-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H44O3 |
| Quantity | 50 mg |
| Synonym | Parigenin Sarsagenin |
| Solubility Information | Ethanol : 1 mg/mL (2.40 mM; Need ultrasonic) |
| SMILES | C[C@@H]1[C@]2(OC[C@@H](C)CC2)O[C@@]3([H])C[C@@]4([H])[C@]5([H])CC[C@]6([H])C[C@@H](O)CC[C@]6(C)[C@@]5([H])CC[C@]4(C)[C@]31[H] |
| Molecular Weight (g/mol) | 416.64 |
| Show More |
Research purposes only