missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SATB2 Antibody (SATB2/7111), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20570-100ug
This item is not returnable.
View return policy
Description
SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SATB2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 | |
| Recombinant fragment (around aa150-350) of human SATB2 protein (exact sequence is proprietary) | |
| 100 μg | |
| Signal Transduction | |
| 23314 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| SATB2/7111 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction