missing translation for 'onlineSavingsMsg'
Learn More

SATB2 Antibody (SATB2/7111), Novus Biologicals™

Mouse Monoclonal Antibody

€ 249.00 - € 494.00

Specifications

Antigen SATB2
Clone SATB2/7111
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18602905
View Documents
Novus Biologicals
NBP3-20570-100ug
100 μg
€ 494.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684864
View Documents
Novus Biologicals
NBP3-20570-20ug
20 μg
€ 249.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

SATB2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Signal Transduction
10mM PBS with 0.05% BSA
23314
IgG2b κ
Protein A or G purified
SATB2/7111
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2
Recombinant fragment (around aa150-350) of human SATB2 protein (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.