Learn More
SBF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-13283
 View more versions of this product
Product Code. 18740624
  • € 624.00 / 0.10mL
Description
SBF2 Polyclonal specifically detects SBF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
SBF2
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.