missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCML2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99032-50ul
This item is not returnable.
View return policy
Description
SCML2 Polyclonal antibody specifically detects SCML2 in Human samples. It is validated for Western Blot
Specifications
| SCML2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| sex comb on midleg (Drosophila)-like 2, sex comb on midleg-like 2 (Drosophila), sex comb on midleg-like protein 2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human SCML2. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10389 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction