missing translation for 'onlineSavingsMsg'
Learn More

SCML2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 227.00 - € 326.00

Specifications

Antigen SCML2
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18646235
View Documents
Novus Biologicals
NBP2-99032-100ul
100 μg
€ 326.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18607735
View Documents
Novus Biologicals
NBP2-99032-50ul
50 μg
€ 227.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SCML2 Polyclonal antibody specifically detects SCML2 in Human samples. It is validated for Western Blot
Specifications

Specifications

SCML2
Western Blot
Unconjugated
Rabbit
Human
sex comb on midleg (Drosophila)-like 2, sex comb on midleg-like 2 (Drosophila), sex comb on midleg-like protein 2
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human SCML2.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
0.2 μm filtered solution in PBS
10389
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado