missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34003-25ul
This item is not returnable.
View return policy
Description
SCN3A Polyclonal specifically detects SCN3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SCN3A | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9NY46 | |
| SCN3A | |
| This antibody was developed against a recombinant protein corresponding to amino acids: NRRKKRRQREHLEGNNKGERDSFPKSESEDSVKRSSFLFSMDGNRLTS | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| brain III voltage-gated sodium channel, KIAA1356, NAC3, Nav1.3, Sodium channel protein brain III subunit alpha, sodium channel protein type 3 subunit alpha, Sodium channel protein type III subunit alpha, sodium channel, voltage-gated, type III, alpha polypeptide, sodium channel, voltage-gated, type III, alpha subunit, Voltage-gated sodium channel subtype III, Voltage-gated sodium channel subunit alpha Nav1.3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6328 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction