Learn More
Ansell™ Scorpio™ 8-352 Series Green Neoprene Immersion Gloves
Product Code. 10452204 Shop All Ansell Products
Click to view available options
Unit Size:
12 pairs
72 pairs
Product Code. 10452204
Brand: Ansell™ 08352 10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Neoprene dipped coating on interlock knit liner for better grip on wet and slippery materials.
  • Flexible
  • Textured finish
  • 100% liquidproof
  • Soft stretchy knit liner with no seams in working area
  • Comfortable to wear, cushions the hand, absorbs perspiration and prevents chafing
  • Good wet grip. Safer handling means increased productivity
  • Easier to manipulate, less tiring to the hand and easier to get on and off
  • Neoprene coating
  • Wide ranging chemical protection
  • Certified FDA21 CFR 177-2600
Applications:
  • Cleaning furnaces, distillation pumps, valves or lines and crackers BTX process
  • Equipment repair and maintenance
  • Filling, blending and charging of raw materials
  • Loading and unloading process equipment
  • Opening and draining pumps valves or lines
  • Opening furnaces draining pumps valves or lines and crackers BTX process
  • Transferring liquids and solids
  • Transferring liquids and solids between vessels and tanks and process equipment
TRUSTED_SUSTAINABILITY
Specifications
Back Material Neoprene
Certifications/Compliance ANSI, EN 388 : 3121, EN 374
Cuff Material Neoprene
Coating Material Neoprene
Laboratory/Cleanroom Use Yes
Product Type Immersion Gloves
Style 8-352
Glove Cuff Style Gauntlet
Lining Interlock Knit Cotton
Palm Type Fully coated-palm
Quantity 72 Pairs
Resistant To Chemicals
Series 8-352