Description
- With PTFE-coated silicone gasket
- Acc. to DIN 168
- Leak-proof with excellent resistance
Specifications
Specifications
| Material | PBT |
| Thread Size (Metric) | GL 25 |
| Liner Material | Silicone Gasket |
| Neck Finish | 14 mm |
| For Use With (Equipment) | Screw-neck sample storage bottles |