missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Screw Vial Caps and Seals

Product Code. 11583860 Shop All Thermo Scientific Products
Click to view available options
Color:
Black
White, Blue
Septum:
Red PTFE/White Silicone
Red PTFE/white silicone seal
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11583860

Brand: Thermo Scientific™ 60180732

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.

Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.

TRUSTED_SUSTAINABILITY

Specifications

Description Blue cap with red PTFE/white silicone seal; Thermo Scientific, PE, Claris, Shimadzu, Varian
Color White, Blue
Material PTFE, Silicone
Septum Red PTFE/white silicone seal
For Use With (Equipment) Thermo Scientific TRACE/AS3000/AI3000/AS800/TriPlus GC, PE Autosystem/AS-2000, Claris 500, ISS-100
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.