missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody (7H12F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41328
This item is not returnable.
View return policy
Description
Seasonal H1N1 Hemagglutinin Monoclonal specifically detects Seasonal H1N1 Hemagglutinin in Influenza A Virus H1N1, Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Seasonal H1N1 Hemagglutinin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA 1 ug/mL | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. Amino Acid Squence: PNIGDQKALTHTEN | |
| 0.1 mg | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| Western Blot, ELISA | |
| 7H12F6 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Influenza virus, Virus | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction