missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody (7H12F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 197.00 - € 509.00
Specifications
| Antigen | Seasonal H1N1 Hemagglutinin |
|---|---|
| Clone | 7H12F6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687548
|
Novus Biologicals
NBP2-41328-0.025mg |
0.025 mg |
€ 197.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18135019
|
Novus Biologicals
NBP2-41328 |
0.1 mg |
€ 509.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Seasonal H1N1 Hemagglutinin Monoclonal specifically detects Seasonal H1N1 Hemagglutinin in Influenza A Virus H1N1, Virus samples. It is validated for Western Blot, ELISA.Specifications
| Seasonal H1N1 Hemagglutinin | |
| 1 mg/mL | |
| Monoclonal | |
| Mouse | |
| Influenza virus, Virus | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| IgG1 | |
| Affinity Purified |
| 7H12F6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. Amino Acid Squence: PNIGDQKALTHTEN | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title