Learn More
SEC63 Mouse, anti-Human, CoraLite Plus 488, Clone: 1C10E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67352-100UL
Description
The SEC63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human antigen. The SEC63 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| SEC63 | |
| 1C10E1 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| ERdj2, PRO2507, SEC63, SEC63 homolog (S. cerevisiae), SEC63L | |
| 100 μL | |
| 11231 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |