Learn More
SEC63 Mouse, anti-Human, CoraLite Plus 488, Clone: 1C10E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67352-100UL
 View more versions of this product
Product Code. 18009395
  • € 450.00 / 100µL
Description
The SEC63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human antigen. The SEC63 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
SEC63
1C10E1
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
ERdj2, PRO2507, SEC63, SEC63 homolog (S. cerevisiae), SEC63L
100 μL
11231
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.