missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC63 Mouse, anti-Human, CoraLite Plus 488, Clone: 1C10E1, Proteintech
Mouse Monoclonal Antibody
€ 140.00 - € 450.00
Specifications
| Antigen | SEC63 |
|---|---|
| Clone | 1C10E1 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18009395
|
Proteintech
CL488-67352-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020646
|
Proteintech
CL488-67352-20UL |
20 μL |
€ 140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SEC63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human antigen. The SEC63 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| SEC63 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 11231 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1C10E1 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| ERdj2, PRO2507, SEC63, SEC63 homolog (S. cerevisiae), SEC63L | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title