missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Xtension Long Nitrile Gloves

Product Code. 15533546
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15533546 Large Pack of 100
15513546 Small Pack of 100
15523546 Medium Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15533546 Supplier Semperit™ Supplier No. 3000004102

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extra-long (300mm) for additional drip protection. Semperit™ Semperguard™ Xtension Long Nitrile Gloves are blue powder-free gloves with rolled cuffs, textured fingers, high comfort and good chemical resistance. Dispenser box = 100 ambidextrous gloves.

  • Extra long: 300mm
  • Extended protection for wrist and forearm
  • Excellent chemical resistance and high wearing comfort
  • Skin friendly: free from phthalates/ softeners and allergenic latex proteins
  • Signal colour blue - suitable for areas subject to HACCP standards
  • Tested and certified according to 1935/2004 - suitable for food contact

Specifications

Certifications/Compliance Conforms to standards EN 420, EN 374, EN455
Classification CE Category III Protective Glove for Complex Risks, CE Class I Examination Glove 
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Grade Certified according to VO 1935/2004 - suitable for contact with food
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Large
Sterility Non-sterile
Thickness (Metric) 0.20 mm
For Use With (Application) Suitable for areas subject to HAACP standards
Glove Cuff Style Beaded/Rolled
Model Xtension Nitrile
Packaging Type Dispenser box
Quantity 100/bx
Length (English) 12 in.
Length (Metric) 300 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.