missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Sensilite™ 48-100 Series White Polyurethane Light Weight Gloves

Product Code. 15222489 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
144 pairs
This item is not returnable. View return policy

Product Code. 15222489

Brand: Ansell™ 48100_6

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for light applications that require good levels of comfort and abrasion resistance.

  • Industrial gloves
  • Abrasion resistance for extended wear life
  • Thin liner and coating offering great dexterity and flexibility
  • Sensilite 48-100 allows extreme tactility where precision is required
  • White colour to ensure process quality and avoid cross contamination
  • Comfort and protection for lightweight applications
  • Offers precise handling of small objects in dry or light oily environments
  • Provides enhanced safety, dexterity and durability

Applications:

  • Assembly and disassembly of equipment
  • Chassis assembly, body trimming
  • Equipment repair and maintenance
  • Loading and unloading rough parts or boxes
  • Picking parts and accessories
  • Production line support and maintenance
TRUSTED_SUSTAINABILITY

Specifications

Back Material Polyurethane
Certifications/Compliance Standard CATEGORY II EN 388 : 4131
Cuff Material Nylon
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Polyurethane
Product Type Lightweight Gloves
Size 6
Glove Cuff Style Knit
Liner Material Nylon
Lining Lined
Palm Type Coated-palm
Quantity 12 Pairs
Resistant To Abrasion resistance
Series 48-100
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.