missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ Manufix™ Sensitive Latex Gloves
Description
Examination and protective gloves
- Single-use
- Non-sterile
- Powder-free
- Natural White
- Natural Rubber Latex
- Shelf life: 3 years
- Store at room temperature, protect from dust, humidity, sunlight, and ozone
- Straight finger
- Ambidextrous fitting
- Rolled rim cuff
- Micro-roughened surface, textured fingers
- Inner glove surface is polymer coated.
Specifications
Specifications
| Certifications/Compliance | DIN EN 455, ASTM F 1671-07 (Viral penetration) and EN 374, EN 420 and PSA 89/686 / EEC, Cat. III |
| Color | Natural White |
| Disposable | Single-use |
| Exterior Finish | Micro-rough |
| Latex Free | No |
| Material | Rubber Latex |
| Palm Thickness (Metric) | ≥0.11 mm |
| Powdered | Powder Free |
| Product Type | Examination Gloves |
| Size | Medium |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?