missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05917-200ul
This item is not returnable.
View return policy
Description
Serpin A11 Polyclonal antibody specifically detects Serpin A11 in Mouse samples. It is validated for ELISA
Specifications
| Serpin A11 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| antiproteinase-like 2, antitrypsin), member 11, member 11, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A11, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Produced in rabbits immunized with purified, recombinant Mouse Serpin A11 (Accession#: NP_955018.2; Met 1-Gly 424) | |
| 200 μg | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 256394 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction