missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 219.00 - € 356.00
Specifications
| Antigen | Serpin A11 |
|---|---|
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682356
|
Novus Biologicals
NBP3-05917-100ul |
100 μg |
€ 219.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625365
|
Novus Biologicals
NBP3-05917-200ul |
200 μg |
€ 275.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602266
|
Novus Biologicals
NBP3-05917-400ul |
400 μg |
€ 356.00
400µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin A11 Polyclonal antibody specifically detects Serpin A11 in Mouse samples. It is validated for ELISASpecifications
| Serpin A11 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| antiproteinase-like 2, antitrypsin), member 11, member 11, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A11, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Produced in rabbits immunized with purified, recombinant Mouse Serpin A11 (Accession#: NP_955018.2; Met 1-Gly 424) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 256394 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title