Learn More
Serpin A6/Cortisol Binding Globulin Antibody (SAIC-42B-10) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20108-0.2mg
 View more versions of this product
Product Code. 18681774
  • € 635.00 / 0.20mg
Description
Serpin A6/Cortisol Binding Globulin Monoclonal antibody specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serpin A6/Cortisol Binding Globulin
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
866
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ
Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry
SAIC-42B-10
Unconjugated
antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin
Peptide 'AQLLQGLGFNLTER' derived from the Corticosteroid-binding globulin SERPINA6, conjugated to KLH.
0.2 mg