missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A6/Cortisol Binding Globulin Antibody (SAIC-42B-10) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 244.00 - € 635.00
Specifications
| Antigen | Serpin A6/Cortisol Binding Globulin |
|---|---|
| Clone | SAIC-42B-10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666014
|
Novus Biologicals
NBP3-20108-0.025mg |
0.025 mg |
€ 244.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681774
|
Novus Biologicals
NBP3-20108-0.2mg |
0.2 mg |
€ 635.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin A6/Cortisol Binding Globulin Monoclonal antibody specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Mass SpectrometrySpecifications
| Serpin A6/Cortisol Binding Globulin | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 866 | |
| IgG κ | |
| Protein A purified |
| SAIC-42B-10 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
| Peptide 'AQLLQGLGFNLTER' derived from the Corticosteroid-binding globulin SERPINA6, conjugated to KLH. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title