missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG3 Heavy Chain Secondary Antibody (5G12cc), Janelia Fluor 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-1599JF549
This item is not returnable.
View return policy
Description
IgG3 Heavy Chain Monoclonal antibody specifically detects IgG3 Heavy Chain in Human samples. It is validated for ELISA
Specifications
| IgG3 Heavy Chain | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG3 from human serum. | |
| 0.2 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| 5G12cc | |
| ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction