missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK3 Antibody (S69), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-47610-0.025mg
This item is not returnable.
View return policy
Description
SHANK3 Monoclonal specifically detects SHANK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications
| SHANK3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100 | |
| DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 85358 | |
| Human | |
| Purified |
| Western Blot | |
| S69-46 | |
| Unconjugated | |
| Q9JLU4 | |
| SHANK3 | |
| Synthetic peptide amino acids 840-857 of rat Shank3 | |
| 0.025 mg | |
| Primary | |
| Detects approx 190kDa. No cross-reactivity against Shank1 or Shank2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction