missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK3 Antibody (S69), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 178.00
Specifications
| Antigen | SHANK3 |
|---|---|
| Clone | S69-46 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot |
Description
SHANK3 Monoclonal specifically detects SHANK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.Specifications
| SHANK3 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2 | |
| SHANK3 | |
| IgG2b | |
| Protein G purified | |
| Detects approx 190kDa. No cross-reactivity against Shank1 or Shank2. |
| S69-46 | |
| Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9JLU4 | |
| 85358 | |
| Synthetic peptide amino acids 840-857 of rat Shank3 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title