missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIELD Scientific SHIELDskin CHEM™ NEO NITRILE™ 300
Click to view available options
Size:
X-Large
Unit Size:
Pack of 400
Description
- Brand signature: Red for chemical risk
- Laboratory gloves for chemical protection
- Neoprene/nitrile gloves
- Non-sterile gloves
- Maximum comfort
- Compliance with the latest European standards
- Category III PPE gloves (Regulation (EU) 2016/425)
- Class 1 MDR (Regulation (EU) 2017/745)
- Extra length from 300 mm (exceeds ISO 21420:2020+A1:2022)
- Biohazard: ISO 374-5:2016
- Biosafety: AQL 0.25 (ISO 374-2:2019 Level 3)
- Virus resistant (ISO 16604:2004 procedure B)
- Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
- Reduced risk of allergies
- Double-walled protection afforded by the twinSHIELD™ technology
- Packaging designed to comply with laboratory environments processes
Specifications
Specifications
| Certifications/Compliance | AQL 0.25 |
| Applications | Laboratory, Industrial |
| Color | Red |
| Finger Thickness (English) | 15.7 mil |
| Finger Thickness (Metric) | 0.40 mm |
| Latex Free | Yes |
| Material | Nitrile, Neoprene |
| Palm Thickness (English) | 12.2 mil |
| Palm Thickness (Metric) | 0.31 mm |
| Size | X-Large |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction