missing translation for 'onlineSavingsMsg'
Learn More

SHIELD Scientific SHIELDskin CHEM™ NEO NITRILE™ 300

Product Code. p-7250954
Encompass_Preferred
Click to view available options
Size:
X-Large
Unit Size:
Pack of 400
This item is not returnable. View return policy

Product Code. 17822254

Brand: SHIELD Scientific 66 9255_10

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

SHIELDskin CHEM™ gloves mark a new level of comfort and protection. SHIELDskin CHEM™ gloves have been designed for laboratory use to protect you from chemical hazards. Using a new CR/NBR blend and the double-dipping process, they protect you from hazards, without compromising comfort.

  • Brand signature: Red for chemical risk
  • Laboratory gloves for chemical protection
  • Neoprene/nitrile gloves
  • Non-sterile gloves
  • Maximum comfort
  • Compliance with the latest European standards
  • Category III PPE gloves (Regulation (EU) 2016/425)
  • Class 1 MDR (Regulation (EU) 2017/745)
  • Extra length from 300 mm (exceeds ISO 21420:2020+A1:2022)
  • Biohazard: ISO 374-5:2016
  • Biosafety: AQL 0.25 (ISO 374-2:2019 Level  3)
  • Virus resistant (ISO 16604:2004 procedure B)
  • Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
  • Reduced risk of allergies
  • Double-walled protection afforded by the twinSHIELD™ technology
  • Packaging designed to comply with laboratory environments processes

Specifications

Certifications/Compliance AQL 0.25
Applications Laboratory, Industrial
Color Red
Finger Thickness (English) 15.7 mil
Finger Thickness (Metric) 0.40 mm
Latex Free Yes
Material Nitrile, Neoprene
Palm Thickness (English) 12.2 mil
Palm Thickness (Metric) 0.31 mm
Size X-Large
Thickness (English) 7.9 mil
Thickness (Metric) 0.20 mm
Glove Cuff Style Beaded
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.