missing translation for 'onlineSavingsMsg'
Learn More

SHIELD Scientific SHIELDskin™ ORANGE NITRILE™ 260

Product Code. p-7250955
Encompass_Preferred
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 900
This item is not returnable. View return policy

Product Code. 17842234

Brand: SHIELD Scientific 67 6231_10

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SHIELDskin™ gloves provide you with a high level of compliance, comfort, and protection. These nitrile/neoprene gloves are specifically designed to protect assays from contamination and protect you from biological and chemical hazards using the revolutionary twinSHIELD™ technology .

  • Brand signature: Orange for biological risk
  • Laboratory gloves for biohazard protection
  • Latex gloves and nitrile/neoprene gloves
  • Non-sterile and sterile gloves
  • Maximum comfort
  • Compliance with the latest European standards
  • Category III PPE gloves (PPE Regulation (EU) 2016/425)
  • Class 1 MDR (Regulation (EU) 2017/745)*
  • Extra length from 260 to 300 mm (meets or exceeds ISO 21420:2020+A1:2022)
  • Biohazard: ISO 374-5:2016
  • Biosafety: AQL 0.25 (ISO 374-2:2019 Level 3)
  • Virus resistant (ISO 16604:2004 Procedure B)
  • Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
  • Reduced risk of allergies
  • Double-walled protection afforded by the twinSHIELD™ technology
  • Packaging designed to comply with laboratory environments processes

Specifications

Certifications/Compliance AQL 0.25
Certifications/Compliance (Europe) PPER (EU) 2016/425, ISO 21420:2020, EN455-2:2015, ISO 374-1:2016-A1:2018
Accelerator Free Yes
Applications Laboratory
AQL 0.25
Color Orange
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 6.7 mil
Finger Thickness (Metric) 0.17 mm
Latex Free Yes
Material Nitrile, Neoprene
Palm Thickness (English) 5.5 mil
Palm Thickness (Metric) 0.14 mm
Powdered Powder Free
Product Type Protective Gloves
Size X-Small
Sterility Non-sterile
Thickness (English) 3.9 mil
Thickness (Metric) 0.10 mm
Length (English) 10.2 in.
Length (Metric) 260 mm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.