missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIELD Scientific SHIELDskin™ ORANGE NITRILE™ 300
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 500
Description
- Brand signature: Orange for biological risk
- Laboratory gloves for biohazard protection
- Latex gloves and nitrile/neoprene gloves
- Non-sterile and sterile gloves
- Maximum comfort
- Compliance with the latest European standards
- Category III PPE gloves (PPE Regulation (EU) 2016/425)
- Extra length from 260 to 300 mm (meets or exceeds ISO 21420:2020+A1:2022)
- Biohazard: ISO 374-5:2016
- Biosafety: AQL 0.25 (ISO 374-2:2019 Level 3)
- Virus resistant (ISO 16604:2004 Procedure B)
- Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
- Reduced risk of allergies
- Double-walled protection afforded by the twinSHIELD™ technology
- Packaging designed to comply with laboratory environments processes
Specifications
Specifications
| Certifications/Compliance | AQL 0.25 |
| Certifications/Compliance (Europe) | PPER (EU) 2016/425, ISO 21420:2020, EN455-2:2015, ISO 374-1:2016-A1:2018 |
| Accelerator Free | Yes |
| Applications | Laboratory |
| AQL | 0.25 |
| Color | Orange |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 6.7 mil |
| Finger Thickness (Metric) | 0.17 mm |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction