missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIP, p Tyr1020 Antibody (SR1167), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21565-100ul
This item is not returnable.
View return policy
Description
SHIP Monoclonal antibody specifically detects SHIP in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
| SHIP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| SR1167 | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:20 | |
| 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
| A synthesized peptide derived from human phospho-SHIP [Tyr1020] (Uniprot #: Q92835) | |
| 100 μg | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 3635 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction