missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIP, p Tyr1020 Antibody (SR1167), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | SHIP |
|---|---|
| Clone | SR1167 |
| Dilution | Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:20 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614255
|
Novus Biologicals
NBP3-21565-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645344
|
Novus Biologicals
NBP3-21565-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHIP Monoclonal antibody specifically detects SHIP in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| SHIP | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:20 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
| A synthesized peptide derived from human phospho-SHIP [Tyr1020] (Uniprot #: Q92835) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1167 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3635 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title