missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT1 Mouse anti-Human, Mouse, Clone: 2.60E+06, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59437-100UL
This item is not returnable.
View return policy
Description
SHMT1 Monoclonal specifically detects SHMT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| SHMT1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:10-1:1043, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| 14 kDa protein, CSHMT, cytoplasmic serine hydroxymethyltransferase, Glycine hydroxymethyltransferase, MGC15229, MGC24556, serine hydroxymethyltransferase 1 (soluble), serine hydroxymethyltransferase, cytosolic, Serine methylase, SHMTEC 2.1.2.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6470 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2600000 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| SHMT1 | |
| Recombinant human SHMT1 (1-483aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction