missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT1 Mouse anti-Human, Mouse, Clone: 2.60E+06, Novus Biologicals™
Mouse Monoclonal Antibody
€ 294.00 - € 486.00
Specifications
| Antigen | SHMT1 |
|---|---|
| Clone | 2600000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 1:10-1:1043, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18271532
|
Novus Biologicals
NBP2-59437-50UL |
50 μL |
€ 294.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18202011
|
Novus Biologicals
NBP2-59437-100UL |
100 μL |
€ 486.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHMT1 Monoclonal specifically detects SHMT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| SHMT1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| 14 kDa protein, CSHMT, cytoplasmic serine hydroxymethyltransferase, Glycine hydroxymethyltransferase, MGC15229, MGC24556, serine hydroxymethyltransferase 1 (soluble), serine hydroxymethyltransferase, cytosolic, Serine methylase, SHMTEC 2.1.2.1 | |
| SHMT1 | |
| IgG1 κ | |
| Protein A purified |
| 2600000 | |
| Western Blot 1:500, Flow Cytometry 1:10-1:1043, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 6470 | |
| Recombinant human SHMT1 (1-483aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title