Learn More
SHOWA™ N-DEX™ Nitrile Laboratory and Industrial Gloves
Description
Provide chemical resistance when exposed to intermittent splashing
- Lightly powdered nitrile with a nontalc vegetable starch
- Ambidextrous
- Single-use
- Length: 9.5 in. (24cm)
- Color: Blue
Intended for occupational safety use only. Not designed for patient contact. Suitable for cleaning and maintaining instruments and equipment.
Specifications
Specifications
| Certifications/Compliance | ASTM F 1671, FDA 21 CFR 170-199, ISO 9002 |
| Color | Blue |
| Disposable | Single-use |
| Hand | Ambidextrous |
| Material | Nitrile |
| Powdered | Powdered |
| Size | Small |
| Thickness (English) | 8 mil |
| Length (English) | 9.5 in. |
| Length (Metric) | 24 cm |