Learn More
SHOWA™ NSK24™ Blue Chemical Resistant Nitrile Coated Gloves
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
Description
Protection from a wide array of solvents, oils and acid.
- Flexibility and dexterity not found in a heavier- mil glove
- Rough-finish texture delivers a sure grip in even the most oily and wet applications
- Interlock liner ensures an added degree of comfort and liner protection
- Color: Blue
Compliance
Data Testing is available upon request.
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 2 |
| Certifications/Compliance | 21 CFR 170-199, ASTM D5526 |
| Certifications/Compliance (Europe) | CE |
| Chlorinated | No |
| Color | Blue |
| Latex Free | Yes |
| Material | Nitrile |
| Size | 10 |
| Thickness (English) | 15 mil |
| Thickness (Metric) | 0.38 mm |
| Show More |