missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ SICCO™ Wall Fastening Kit for Desiccators
Click to view available options
Material:
Aluminum
Unit Size:
Each
Description
- For safe fastening or hanging of desiccators on the wall
- Consists of one pair of wall rails, one pair of fastening rails, six cylinder screws and one hexagon wrench
- Screws for fixation of the rails on your wall are not included
Specifications
Specifications
| Material | Aluminum |
| Height (Metric) | 10 mm |
| Height (English) | 0.4 in. |
| Weight (English) | 1.4 lb. |
| Weight (Metric) | 650 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction