Learn More
Silica G-TLC, Contains Gypsum, MP
Product Code. 17837054 Shop All MP Biomedicals Products
Click to view available options
:
MP Silica G-TLC, 25 kg
Product Code. 17837054
Brand: MP Biomedicals 0204674
Available with or without binder and with or without fluorescent indicators
Produced from the same highly standardized raw material as the MP EcoChrom™ Silica for Column Chromatography
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 2230°C
Melting Point 1710°C
CAS 7631-86-9
Color White
pH 6.5
Physical Form Powder
Quantity 25 kg
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Density 0.4 g/mL
Chemical Name or Material Silicon dioxide
Show More Show Less
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.